Merck
  • Echinacea purpurea root extract enhances the adipocyte differentiation of 3T3-L1 cells.
Echinacea purpurea root extract enhances the adipocyte differentiation of 3T3-L1 cells.
Archives of pharmacal research (2013-10-03)
Dong-Mi Shin, Kyeong-Mi Choi, Youn-Sun Lee, Wonkyun Kim, Kyong-Oh Shin, Seikwan Oh, Jae-Chul Jung, Mi Kyeong Lee, Yong-Moon Lee, Jin Tae Hong, Yeo-Pyo Yun, Hwan-Soo Yoo
ABSTRACT
Echinacea purpurea has been shown to have anti-diabetic activities; for example, it activates peroxisome proliferator-activated receptor γ (PPARγ) and increases insulin-stimulated glucose uptake. Adipogenesis has been used to study the insulin signaling pathway and to screen anti-diabetic compounds. The present study was conducted to investigate the effects of an ethanol extract of E. purpurea (EEEP) and its constituents on the insulin-induced adipocyte differentiation of 3T3-L1 preadipocytes. When adipocyte differentiation was induced with insulin plus 3-isobutyl-1-methylxanthine and dexamethasone, the accumulation of lipid droplets and the cellular triglyceride content were significantly increased by EEEP. The expressions of PPARγ and C/EBPα in adipocytes treated with EEEP were gradually increased as compared with control cells. Fat accumulation and triglyceride content of adipocytes treated with dodeca-2(E),4(E)-dienoic acid isobutylamide were significantly increased as compared with control cells. The expressions of PPARγ and C/EBPα in adipocytes treated with dodeca-2(E),4(E)-dienoic acid isobutylamide were significantly higher than in control cells. These results suggest EEEP promotes the adipogenesis that is partially induced by insulin and that dodeca-2(E),4(E)-dienoic acid isobutylamide appears to be responsible for EEEP-enhanced adipocyte differentiation.
Product Description
Chlorogenic acid, primary reference standard
Sigma-Aldrich
Troglitazone, ≥98% (HPLC)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Echinacoside, analytical standard
Sigma-Aldrich
Echinacoside, 98% (HPLC)
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard