Merck
  • miR-21 regulates triglyceride and cholesterol metabolism in non-alcoholic fatty liver disease by targeting HMGCR.
miR-21 regulates triglyceride and cholesterol metabolism in non-alcoholic fatty liver disease by targeting HMGCR.
International journal of molecular medicine (2015-01-22)
Chuanzheng Sun, Feizhou Huang, Xunyang Liu, Xuefei Xiao, Mingshi Yang, Gui Hu, Huaizheng Liu, Liangkan Liao
ABSTRACT
Non-alcoholic fatty liver disease (NAFLD) has emerged as a public health issue with a prevalence of 15-30% in Western populations and 6-25% in Asian populations. Certain studies have revealed the alteration of microRNA (miRNA or miR) profiles in NAFLD and it has been suggested that miR-21 is associated with NAFLD. In the present study, we measured the serum levels of miR-21 in patients with NAFLD and also performed in vitro experiments using a cellular model of NAFLD to further investigate the effects of miR-21 on triglyceride and cholesterol metabolism. Furthermore, a novel target through which miR-21 exerts its effects on NAFLD was identified. The results revealed that the serum levels of miR-21 were lower in patients with NAFLD compared with the healthy controls. In addition, 3-hydroxy-3-methylglutaryl-co-enzyme A reductase (HMGCR) expression was increased in the serum of patients with NAFLD both at the mRNA and protein level. To mimic the NAFLD condition in vitro, HepG2 cells were treated with palmitic acid (PA) and oleic acid (OA). Consistent with the results obtained in the in vivo experiments, the expression levels of miR-21 were decreased and those of HMGCR were increased in the in vitro model of NAFLD. Luciferase reporter assay revealed that HMGCR was a direct target of miR-21 and that miR-21 exerted an effect on both HMGCR transcript degradation and protein translation. Furthermore, the results from the in vitro experiments revealed that miR-21 decreased the levels of triglycerides (TG), free cholesterol (FC) and total cholesterol (TC) in the PA/OA-treated HepG2 cells and that this effect was attenuated by HMGCR overexpression. Taken together, to the best of our knowledge, the present study is the first to report that miR-21 regulates triglyceride and cholesterol metabolism in an in vitro model of NAFLD, and that this effect is achieved by the inhibition of HMGCR expression. We speculate that miR-21 may be a useful biomarker for the diagnosis and treatment of NAFLD.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
USP
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Palmitic acid, analytical standard
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)