Merck
  • A new route for synthesis of silver:gold alloy nanoparticles loaded within phosphatidylcholine liposome structure as an effective antibacterial agent against Pseudomonas aeruginosa.
A new route for synthesis of silver:gold alloy nanoparticles loaded within phosphatidylcholine liposome structure as an effective antibacterial agent against Pseudomonas aeruginosa.
Journal of liposome research (2014-06-25)
Amir H Salehi, Majid Montazer, Tayebeh Toliyat, Mahnaz Mahmoudi-Rad
ABSTRACT
Ag:Au alloy nanoparticles were successfully synthesized through the new route using co-reduction method with silver nitrate, chloroauric acid, cetyl trimethyl ammonium bromide (CTAB) and sodium borohydride at room temperature. The Ag:Au alloy nanoparticles were then loaded within the phosphatidylcholine (97%) liposome structure. Various molar ratios of phosphotidylcholine and CTAB to the total metals were investigated showing its importance on the stability of nanocomposites suspension. The size distribution and morphology of encapsulated nanoparticles within the liposome structure were studied via ultraviolet (UV)-visible spectrum, transmission electron microscope (TEM) micrographs, and dynamic light scattering data. The synthesis of alloy nanoparticles were confirmed with formation of single band at 430, 465 and 500 nm for 75:25, 50:50 and 25:75 Ag:Au mole ratios, respectively. The TEM micrographs of different samples indicated formation of three various nanocomposite structures with size of 82-300 nm. The antibacterial activities of Ag:Au nanocomposites were studied against Pseudomonas aeruginosa through well-diffusion agar. The minimum inhibitory concentration (MIC) and minimum bactericidal concentration (MBC) were determined by Broth microdilution method. The results showed that 10 ppm nanocomposite reasonably killed the above bacteria.
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%