Merck
  • Lung transplantation in chronic obstructive pulmonary disease: long-term survival, freedom from bronchiolitis obliterans syndrome, and factors influencing outcome.
Lung transplantation in chronic obstructive pulmonary disease: long-term survival, freedom from bronchiolitis obliterans syndrome, and factors influencing outcome.
Clinical transplantation (2015-02-11)
Mohamed Zeriouh, Prashant N Mohite, Anton Sabashnikov, Bartlomiej Zych, Nikhil P Patil, Diana Garcia-Saez, Achim Koch, Ali Ghodsizad, Alexander Weymann, Simona Soresi, Thorsten Wittwer, Yeong-Hoon Choi, Jens Wippermann, Thorsten Wahlers, Aron-Frederik Popov, André R Simon
ABSTRACT
Lung transplantation (LTx) remains the definitive treatment for end-stage lung failure, whereas chronic obstructive pulmonary disease (COPD) represents one of the main diagnoses leading to the indication for a transplant. We sought to assess long-term outcomes after LTx in patients diagnosed with COPD and analyze factors influencing outcome in this frequent patient cohort. Between January 2007 and November 2013, a total of 88 LTx were performed in patients with COPD in our institution. Patients with emphysema associated with alpha1-antitrypsin deficiency were excluded from this observation. The study design was a retrospective review of the prospectively collected data. A large number of pre-, intra-, and postoperative variables were analyzed including long-term survival and freedom from bronchiolitis obliterans syndrome (BOS). Furthermore, impact of different variables on survival was analyzed. Preoperative donor data indicated a large proportion of marginal donors. While the overall cumulative survival after six yr was 57.4%, the results in terms of BOS-free survival in long-term follow-up were 39.7% after six yr. Patients with COPD were also associated with a low incidence (2.3%) of the need for postoperative extracorporeal life support (ECLS). Long-term results after LTx in patients with COPD are acceptable with excellent survival, freedom from BOS, and low use of ECLS postoperatively despite permanently increasing proportion of marginal organs used.
Product Description
Trometamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
SAFC
Tromethamine
Sigma-Aldrich
Potassium hydride, in paraffin
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O