Merck
  • Process development for biological production of butanol from Eastern redcedar.
Process development for biological production of butanol from Eastern redcedar.
Bioresource technology (2014-12-03)
Kan Liu, Hasan K Atiyeh, Oscar Pardo-Planas, Karthikeyan D Ramachandriya, Mark R Wilkins, Thaddeus C Ezeji, Victor Ujor, Ralph S Tanner
ABSTRACT
Eastern redcedar is an invasive softwood species in Oklahoma and across grasslands in the Central Plains of the United States and potential feedstock for butanol production. Butanol has higher energy content than ethanol and can be upgraded to jet and diesel fuels. The objective of this study was to develop a process for production of butanol from redcedar. Results showed that Clostridium acetobutylicum ATCC 824 and Clostridium beijerinckii NCIMB 8052 did not grow in fermentation medium with citrate buffer. However, both strains grew in the medium with acetate buffer, resulting in 3-4g/L greater butanol than without acetate. Detoxification of redcedar hydrolyzate was required to increase butanol concentration from 1 to 13g/L. Hydrolyzate was detoxified by activated carbon to remove inhibitors. Fermentations in detoxified redcedar hydrolyzate reached 13g/L butanol and 19g/L total ABE, comparable to glucose control. This shows the potential for redcedar use in butanol production.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Levulinic acid, 98%
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Levulinic acid, ≥97%, FG
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Levulinic acid, natural, 99%, FG
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Supelco
Levulinic acid, analytical standard
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%