Merck
  • Differentiation of South American crack and domestic (US) crack cocaine via headspace-gas chromatography/mass spectrometry.
Differentiation of South American crack and domestic (US) crack cocaine via headspace-gas chromatography/mass spectrometry.
Drug testing and analysis (2014-10-11)
Valerie L Colley, John F Casale
ABSTRACT
South American 'crack' cocaine, produced directly from coca leaf, can be distinguished from US domestically produced crack on the basis of occluded solvent profiles. In addition, analysis of domestically produced crack indicates the solvents that were used for cocaine hydrochloride (HCl) processing in South America. Samples of cocaine base (N=3) from South America and cocaine from the USA (N=157 base, N=88 HCl) were analyzed by headspace-gas chromatography-mass spectrometry (HS-GC-MS) to determine their solvent profiles. Each cocaine HCl sample was then converted to crack cocaine using the traditional crack production method and re-examined by HS-GC-MS. The resulting occluded solvent profiles were then compared to their original HCl solvent profiles. Analysis of the corresponding crack samples confirmed the same primary processing solvents found in the original HCl samples, but at reduced levels. Domestically seized crack samples also contained reduced levels of base-to-HCl conversion solvents. In contrast, analysis of South American crack samples confirmed the presence of low to high boiling hydrocarbons and no base-to-HCl conversion solvents. The presented study showed analysis of crack cocaine samples provides data on which processing solvents were originally utilized in the production of cocaine HCl in South America, prior to conversion to crack cocaine. Determination of processing solvents provides valuable information to the counter-drug intelligence community and assists the law enforcement community in determining cocaine distribution and trafficking routes throughout the world.
Product Description
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone-d6, 99.5 atom % D
Sigma-Aldrich
Acetone-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Supelco
Isobutyl acetate, analytical standard
Sigma-Aldrich
Acetone-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
2-Propanol-d8, 99.5 atom % D
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetone-d6, "100%", 99.95 atom % D
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
2-Chloro-2-methylpropane-d9, 99 atom % D
Sigma-Aldrich
Acetone-d6, 99.9 atom % D
Sigma-Aldrich
Isobutyl acetate, FCC, FG
Sigma-Aldrich
Acetone-d6, 99.9 atom % D
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Isobutyl acetate, natural, ≥97%, FCC, FG
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer