Merck
  • BMP-2 and -4 produced by vascular smooth muscle cells from atherosclerotic lesions induce monocyte chemotaxis through direct BMPRII activation.
BMP-2 and -4 produced by vascular smooth muscle cells from atherosclerotic lesions induce monocyte chemotaxis through direct BMPRII activation.
Atherosclerosis (2014-05-13)
Alex Yuri Simões Sato, Guilherme Linhares Bub, Alexandre Holthausen Campos
ABSTRACT
Monocytes and macrophages, together with vascular smooth muscle cells (VSMCs), play key roles at all stages of atherogenesis. There is also growing evidence that BMP signaling is involved in vascular diseases, including atherosclerosis. Here we evaluate the role played by the BMP agonist/antagonist axis in monocyte recruitment during atherogenesis. Using ApoE-/- mice and BMPs, Gremlin and BMPRII siRNAs we show that BMPs (2 and 4) and their antagonist Gremlin are co-expressed in murine and human atherosclerotic vessels. Additionally, those genes are co-expressed and upregulated in cultured vascular smooth muscle cells early in atherosclerosis formation in ApoE-/- mice. Furthermore, we demonstrate that BMP-2 and -4 produced in atherosclerotic VSMCs promote, whereas Gremlin inhibits, monocyte chemoattraction. Finally, we demonstrate that chemotaxis induction occurs through direct BMP receptor II (BMPRII) activation. These findings suggest that the balance between BMPs (2 and 4) and Gremlin levels modulate crosstalk processes between vascular and immune cells and ultimately the homeostasis in normal vasculature. They also indicate that under pro-atherogenic conditions, BMP signaling prevails, favoring monocyte recruitment and inflammation. Manipulation of BMP signaling may enable the identification of novel molecular approaches for preventing, stabilizing, and reverting atherosclerosis.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Bmpr2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Bmp4
Sigma-Aldrich
Anti-BMP4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human BMPR2
Sigma-Aldrich
MISSION® esiRNA, targeting human BMP2
Sigma-Aldrich
MISSION® esiRNA, targeting human BMP4
Sigma-Aldrich
Methyl Green, zinc chloride salt, for microscopy (Bact., Bot., Hist.)
Sigma-Aldrich
Methyl Green, zinc chloride salt, certified by the BSC, Dye content 85 %
Sigma-Aldrich
Methyl Green, zinc chloride salt, <0.5% crystal violet, Dye content 80 %
Sigma-Aldrich
Methyl Green, zinc chloride salt, ~85%