Merck
  • Dispersion of titanate nanotubes for nanomedicine: comparison of PEI and PEG nanohybrids.
Dispersion of titanate nanotubes for nanomedicine: comparison of PEI and PEG nanohybrids.
Dalton transactions (Cambridge, England : 2003) (2014-11-20)
Anne-Laure Papa, Julien Boudon, Vanessa Bellat, Alexis Loiseau, Harender Bisht, Fadoua Sallem, Rémi Chassagnon, Véronique Bérard, Nadine Millot
ABSTRACT
In the present study, we report the dispersion of titanate nanotubes (TiONts) via polymer grafting (PolyEthylene Glycol, PEG) or polymer adsorption (polyethylene imine, PEI) where different TiONts/polymer ratios have been investigated. The TiONts/PEI and TiONts/PEG nanohybrids were characterized by scanning and transmission electron microscopy as well as by zeta potential measurements in order to determine both their dispersion state and stability in water (at different pH for zetametry). The nature of the chemical bonds at the surface of these nanohybrids was investigated by Fourier-transformed infrared (FTIR) spectroscopy while the grafting densities of PEG on the nanotubes were quantified by thermogravimetric analyses (TGA). The nanohybrids reported here are promising tools for biotechnology applications due to their tubular morphology, their very good dispersion in water and the reactivity of their surface.
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Dibutyltin dilaurate, 95%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets