Merck
  • Controlled release of liposome-encapsulated Naproxen from core-sheath electrospun nanofibers.
Controlled release of liposome-encapsulated Naproxen from core-sheath electrospun nanofibers.
Carbohydrate polymers (2014-07-20)
Zhuang Li, Hongliang Kang, Ning Che, Zhijing Liu, Pingping Li, Weiwei Li, Chao Zhang, Chun Cao, Ruigang Liu, Yong Huang
ABSTRACT
Naproxen (NAP) loaded nanofibers of different structures have been successfully prepared by electrospinning. The structures of the nanofibers are NAP and cellulose acetate (CA) mixed nanofibers (NF-1), nanofibers with NAP/CA mixed core and CA sheath (NF-2), and NAP loaded liposomes and sodium hyaluronate (HA-Na) mixed core with CA sheath (NF-3). The structure and morphology of the nanofibers were characterized and the drug release behaviors were investigated. It was found that NAP can disperse in the HA-Na or CA matrix in molecular level without formation of NAP crystals and dimers. The drug release behaviors of NF-1 and NF-2 show a non-Fickian diffusion mechanism, while the NF-3 shows a specific drug release behavior with a burst release within 8h followed by a sustained drug release for 12 days. The particular two-stage drug release behavior of NF-3 nanofibers offers the materials promising applications as wound dressing materials.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Naproxen, European Pharmacopoeia (EP) Reference Standard
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Supelco
Naproxen, VETRANAL®, analytical standard
Sigma-Aldrich
Naproxen, meets USP testing specifications
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
USP
Naproxen, United States Pharmacopeia (USP) Reference Standard