Merck
  • Kinetic studies of inverse electron demand Diels-Alder reactions (iEDDA) of norbornenes and 3,6-dipyridin-2-yl-1,2,4,5-tetrazine.
Kinetic studies of inverse electron demand Diels-Alder reactions (iEDDA) of norbornenes and 3,6-dipyridin-2-yl-1,2,4,5-tetrazine.
Tetrahedron letters (2014-08-26)
Astrid-Caroline Knall, Manuel Hollauf, Christian Slugovc
ABSTRACT
Inverse electron demand Diels-Alder additions (iEDDA) between 1,2,4,5-tetrazines and olefins have recently found widespread application as a novel 'click chemistry' scheme and as a mild technique for the modification of materials. Norbornenes are, due to their straightforward synthetic availability, especially interesting in the latter context. Therefore, the reactivity of different norbornene-based compounds was compared with unsubstituted norbornene and other alkenes using UV-vis measurements for the determination of reaction rates under pseudo first order conditions. Thereby,
Product Description
Supelco
1-Hexene, analytical standard
Sigma-Aldrich
1-Hexene, 97%
Sigma-Aldrich
Dicyclopentadiene, contains BHT as stabilizer
Sigma-Aldrich
1-Hexene, ≥99%
Sigma-Aldrich
1-Hexene, 97%
Supelco
Styrene, analytical standard
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Sigma-Aldrich
Cyclopentene, 96%