Merck
  • High-throughput chemical modification of oligonucleotides for systematic structure-activity relationship evaluation.
High-throughput chemical modification of oligonucleotides for systematic structure-activity relationship evaluation.
Bioconjugate chemistry (2014-11-15)
Daniel Zewge, Francis Gosselin, Denise M Kenski, Jenny Li, Vasant Jadhav, Yu Yuan, Sandhya S Nerurkar, David M Tellers, W Michael Flanagan, Ian W Davies
ABSTRACT
Chemical modification of siRNA is achieved in a high-throughput manner (96-well plate format) by copper catalyzed azide-alkyne cycloadditions. This transformation can be performed in one synthetic operation at up to four positions with complete specificity, good yield, and acceptable purity. As demonstrated here, this approach extends the current synthetic options for oligonucleotide modifications and simultaneously facilitates the systematic, rapid biological evaluation of modified siRNA.
Product Description
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Supelco
Dichloromethane, Selectophore, for ion-selective electrodes, ≥99.5%
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
2,6-Dimethylpyridine, ≥99%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
N,N-Dimethylacetamide, analytical standard
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
N,N-Dimethylacetamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Citric acid, 99%
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylacetamide-d9, 99 atom % D
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
N,N-Dimethylacetamide, suitable for peptide synthesis, ≥99.8% (GC)
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer