Merck
  • A truncation variant of the cation channel P2RX5 is upregulated during T cell activation.
A truncation variant of the cation channel P2RX5 is upregulated during T cell activation.
PloS one (2014-09-03)
Pierre Abramowski, Christoph Ogrodowczyk, Roland Martin, Olaf Pongs
ABSTRACT
Members of the P2X family of ligand-gated cation channels (P2RX) are expressed by various cell types including neurons, smooth- and cardiac muscle cells, and leukocytes. The channels mediate signalling in response to extracellular ATP. Seven subunit isoforms (P2RX1-P2RX7) have been identified and these can assemble as homo- and heterotrimeric molecules. In humans, P2RX5 exists as a natural deletion mutant lacking amino acids 328-349 of exon 10, which are part of transmembrane (TM) 2 and pre-TM2 regions in other organisms like rat, chicken and zebrafish. We show that P2RX5 gene expression of human T lymphocytes is upregulated during activation. P2RX5 is recruited to the cell surface. P2RX5-siRNA-transfected CD4+ T cells produced twofold more IL-10 than controls. Surface and intracellular P2RX5 expression was upregulated in activated antigen-specific CD4+ T cell clones. These data indicate a functional role of the human P2RX5 splice variant in T cell activation and immunoregulation.
Product Description
SAFC
BIS-TRIS
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
SAFC
BIS-TRIS
Sigma-Aldrich
Monoclonal Anti-P2RX5 antibody produced in mouse, clone 1C5, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-P2RX5 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Nitrogen, ≥99.998%