Merck
  • A concomitant loss of dormant origins and FANCC exacerbates genome instability by impairing DNA replication fork progression.
A concomitant loss of dormant origins and FANCC exacerbates genome instability by impairing DNA replication fork progression.
Nucleic acids research (2014-03-05)
Spencer W Luebben, Tsuyoshi Kawabata, Charles S Johnson, M Gerard O'Sullivan, Naoko Shima
ABSTRACT
Accumulating evidence suggests that dormant DNA replication origins play an important role in the recovery of stalled forks. However, their functional interactions with other fork recovery mechanisms have not been tested. We previously reported intrinsic activation of the Fanconi anemia (FA) pathway in a tumor-prone mouse model (Mcm4chaos3) with a 60% loss of dormant origins. To understand this further, we introduced a null allele of Fancc (Fancc-), encoding a member of the FA core complex, into the Mcm4chaos3 background. Primary embryonic fibroblasts double homozygous for Mcm4chaos3 and Fancc- (Mcm4chaos3/chaos3;Fancc-/-) showed significantly increased levels of markers of stalled/collapsed forks compared to either single homozygote. Interestingly, a loss of dormant origins also increased the number of sites in which replication was delayed until prophase, regardless of FA pathway activation. These replication defects coincided with substantially elevated levels of genome instability in Mcm4chaos3/chaos3;Fancc-/- cells, resulting in a high rate of perinatal lethality of Mcm4chaos3/chaos3;Fancc-/- mice and the accelerated tumorigenesis of surviving mice. Together, these findings uncover a specialized role of dormant origins in replication completion while also identifying important functional overlaps between dormant origins and the FA pathway in maintaining fork progression, genome stability, normal development and tumor suppression.
Product Description
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
Supelco
Digoxigenin, analytical standard
Roche
Anti-Digoxigenin-Rhodamine, Fab fragments, from sheep
Roche
Anti-Digoxigenin, from mouse IgG1κ (clone 1.71.256)
Sigma-Aldrich
Goat anti-MCM4 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.