Merck
  • Decreased expression of heat shock protein 20 in colorectal cancer and its implication in tumorigenesis.
Decreased expression of heat shock protein 20 in colorectal cancer and its implication in tumorigenesis.
Journal of cellular biochemistry (2014-09-05)
Young-Tae Ju, Seung-Jin Kwag, Hee Jin Park, Eun-Jung Jung, Chi-Young Jeong, Sang-Ho Jeong, Young-Joon Lee, Sang-Kyung Choi, Kee Ryeon Kang, Young-Sool Hah, Soon-Chan Hong
ABSTRACT
Heat shock protein 20 (HSP20), which is a member of the small heat shock protein family, is known to participate in many pathological processes, such as asthma, intimal hyperplasia, and insulin resistance. However, the function of HSP20 in cancer development is not yet fully understood. In this study, we identified HSP20 as a down-regulated protein in 20 resected colorectal cancer (CRC) specimens compared with their paired normal tissues. Because HSP20 proteins were barely detectable in HCT-116 cells (a human colorectal cancer cell line), recombinant adenovirus encoding HSP20 (Ad-HSP20) was used to induce HSP20 overexpression in HCT-116 cells. Infection of Ad-HSP20, but not control adenovirus (Ad-GFP), reduced viability, and induced massive apoptosis in a time-dependent manner. The forced expression of HSP20 enhanced caspase-3/7 activity and down-regulated the anti-apoptotic Bcl-xL and Bcl-2 mRNA and protein levels. In addition, immunohistochemical analysis of 94 CRC specimens for HSP20 protein showed that reduced HSP20 expression was related to advanced TNM stage, lymph node metastasis, and tumor recurrence. Our study shows, for the first time, that expression of the HSP20 protein has a pro-death role in colorectal cancer cells. Therefore, HSP20 may have value as a prognostic tumor marker and its overexpression might be a novel strategy for CRC therapy.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%