Merck
  • Characterization of complex lipid mixtures in contaminant exposed JEG-3 cells using liquid chromatography and high-resolution mass spectrometry.
Characterization of complex lipid mixtures in contaminant exposed JEG-3 cells using liquid chromatography and high-resolution mass spectrometry.
Environmental science and pollution research international (2014-06-28)
Eva Gorrochategui, Josefina Casas, Elisabet Pérez-Albaladejo, Olga Jáuregui, Cinta Porte, Sílvia Lacorte
ABSTRACT
The aim of this study was to develop a method based on ultra high performance liquid chromatography coupled with mass spectrometry (UHPLC-MS) for lipid profiling in human placental choriocarcinoma (JEG-3) cells. Lipids were solid-liquid extracted from JEG-3 cells using a solution of chloroform/methanol (2:1, v/v) in a simple procedure requiring minimal sample alteration. Simultaneous separation of complex lipid mixtures in their major classes was achieved with a reversed-phase (C8) UHPLC column and a mobile phase containing methanol with 1 mM ammonium formate and 0.2 % formic acid (A)/water with 2 mM ammonium formate and 0.2 % formic acid (B). Lipids were characterized using time-of-flight (TOF) and Orbitrap under full scan and positive electrospray ionization mode with both analyzers. A total of 178 species of lipids, including 37 phosphatidylcholines (PC), 32 plasmalogen PC, 9 lyso PC, 4 lyso plasmalogen PC, 30 triacylglycerols, 22 diacylglycerols, 7 cholesterol esters, 25 phosphatidylethanolamines, and 12 sphingomyelins, were identified using TOF and Orbitrap. The identification of all lipid classes was based on exact mass characterization with an error < 5 ppm. The developed methodology was applied to study lipid alterations in human placental cells against the exposure to perfluorinated chemicals (PFCs) and tributyltin (TBT).
Product Description
Supelco
Heptadecafluorooctanesulfonic acid solution, 100 μg/mL in methanol, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Heptafluorobutyric acid solution, 0.5 M in H2O, LiChropur, suitable for ion pair chromatography
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Sigma-Aldrich
Undecafluorohexanoic acid, ≥97.0% (T)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Pentadecafluorooctanoic acid solution, 100 μg/mL in methanol, analytical standard