Merck
  • Phenolic compounds involved in grafting incompatibility of Vitis spp: development and validation of an analytical method for their quantification.
Phenolic compounds involved in grafting incompatibility of Vitis spp: development and validation of an analytical method for their quantification.
Phytochemical analysis : PCA (2014-06-04)
Sara Canas, Maria Assunção, João Brazão, Geni Zanol, José Eduardo Eiras-Dias
ABSTRACT
Graft incompatibility of Vitis spp is an unresolved worldwide problem with important economic consequences. Grafting comprises a complex set of morphological and physiological alterations, in which the phenolic compounds seem to be strongly involved. Therefore, a detailed analysis and recognition of structural phenolic compounds diversity in the two partners of a Vitis graft is of great importance to evaluate their role as markers of graft establishment. To optimise a sample extraction method, and to develop and validate a high-performance liquid chromatography (HPLC) method for the simultaneous determination of phenolic acids and flavonols in the graft union so as to understand their behaviour in the metabolism of the scion-rootstock system, using compatible and incompatible combinations of a Syrah cultivar and two rootstocks (R110 and SO4). Sixty extracts of Vitis grafting tissues were prepared and analysed by HPLC for the qualitative and quantitative determination of their phenolic profile. Among the phenolic compounds identified in the samples, one benzoic acid (gallic acid), three cinnamic acids (caffeic acid, ferulic acid and sinapic acid) and two flavonols (catechin and epicatechin) are potentially suitable as markers of graft incompatibility. The method developed presents good performance and lends itself readily for application in routine analysis of the phenolic composition of Vitis grafting tissues to distinguish compatible and incompatible combinations in the graft callusing stage.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Gallic acid monohydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.0% (T)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.5%, Ultra pure
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Supelco
Acetone, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free