Merck
  • Oral L-carnitine supplementation increases trimethylamine-N-oxide but reduces markers of vascular injury in hemodialysis patients.
Oral L-carnitine supplementation increases trimethylamine-N-oxide but reduces markers of vascular injury in hemodialysis patients.
Journal of cardiovascular pharmacology (2015-01-31)
Kei Fukami, Sho-ichi Yamagishi, Kazuko Sakai, Yusuke Kaida, Miyuki Yokoro, Seiji Ueda, Yoshifumi Wada, Masayoshi Takeuchi, Makiko Shimizu, Hiroshi Yamazaki, Seiya Okuda
ABSTRACT
Food or supplement-derived L-carnitine is changed to trimethylamine (TMA) by interstinal microbiota, which is further metabolized to trimethylamine-N-oxide (TMAO), being involved in the promotion of atherosclerosis in animal models. Meanwhile, carnitine deficiency has played a role in accelerated atherosclerosis in hemodialysis (HD) patients. However, effects of oral L-carnitine supplementation on circulating levels of TMAO and markers of vascular injury and oxidative stress in patients on HD remain unclear. In this study, we addressed the issue. Thirty-one HD patients with carnitine deficiency were treated with oral L-carnitine (900 mg/d) for 6 months. At baseline and after treatment, clinical variables including circulating levels of carnitine fractions, TMA, TMAO, advanced glycation end products (AGE), soluble forms of intracellular adhesion molecule-1 (sICAM-1), vascular cell adhesion molecule-1 (sVCAM-1), and malondialdehyde (MDA) were measured. Oral L-carnitine supplementation significantly increased total, free, acyl carnitine, and plasma TMA and TMAO levels, whereas it decreased markers of vascular injury and oxidative stress such as sICAM-1, sVCAM-1, and MDA levels. TMA and TMAO levels at baseline were correlated with each other, and free carnitine was independently associated with TMAO levels. Furthermore, change in AGE values from baseline ([INCREMENT]AGE) was positively correlated with [INCREMENT]sICAM-1 (P = 0.043) and was a sole independent determinant of [INCREMENT]sICAM-1 (R = 0.133, P = 0.043). This study demonstrated that although oral L-carnitine supplementation was associated with increased TMAO levels, it might be beneficial on vascular injury in patients on HD. Vasculoprotective properties of L-carnitine supplementation in HD patients might be ascribed partly to its inhibitory actions on AGE.
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
4,4′-Diaminodiphenylmethane, analytical standard
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Uric acid, ≥99%, crystalline
SAFC
Cholesterol, Plant-Derived, SyntheChol®