Merck
  • Differential role of MACC1 expression and its regulation of the HGF/c‑Met pathway between breast and colorectal cancer.
Differential role of MACC1 expression and its regulation of the HGF/c‑Met pathway between breast and colorectal cancer.
International journal of oncology (2015-03-05)
Aiko Sueta, Yutaka Yamamoto, Mutsuko Yamamoto-Ibusuki, Mitsuhiro Hayashi, Takashi Takeshita, Satoko Yamamoto, Yoko Omoto, Hirotaka Iwase
ABSTRACT
The newly identified gene, metastasis‑associated in colon cancer 1 (MACC1), is suggested to be a transcriptional regulator of c‑Met, leading to cancer progression in colorectal cancer. To date however, little is known of the role of MACC1 in breast cancer. In a series of 300 breast cancer patients, we analyzed the association of MACC1 mRNA and protein expression with breast cancer survival using Cox proportional hazard models. In an in vitro study, we evaluated activities of c‑Met protein after transfection with a MACC1‑harboring plasmid as well as the binding ability of MACC1 to the c‑Met promoter using a chromatin immunoprecipitation (ChIP) assay. In survival analyses, reduced MACC1 expression was associated with patient mortality. MACC1 expression was an independent prognostic factor in multivariate analysis. In the cell lines tested, MACC1 expression was much higher in colorectal than in breast cancer cells. After cells were transfected with MACC1, c‑Met expression was not induced in MCF7 cells, whereas corresponding c‑Met expression was upregulated in SW480 cells. Further, SW480 cells transfected with MACC1 showed enhanced migratory ability, whereas in MDA‑MB‑231 cells, transfection of MACC1 had no impact on this ability. In ChIP assay, the binding of MACC1 to the c‑Met promoter was suggested in SW480 cells, but not in MCF7 cells. In conclusion, our findings provide some novel insights into the role of MACC1 in breast cancer, indicating that it plays different roles in breast and several other cancers. There is a possibility that MACC1 does not modulate the transcriptional role of c‑Met signaling in breast cancer.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Macc1
Sigma-Aldrich
MISSION® esiRNA, targeting human MACC1
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O