Merck
  • Stereoelectronic effects: a simple yet powerful tool to manipulate anion affinity.
Stereoelectronic effects: a simple yet powerful tool to manipulate anion affinity.
Organic & biomolecular chemistry (2014-12-24)
Masoud Samet, Alireza Fattahi, Steven R Kass
ABSTRACT
Different strategies are employed in designing strong and selective anion receptors but stereoelectronic effects have been largely ignored. In this work, the stereo configuration of a non-interacting ether is found to have a large impact of more than two orders of magnitude on the binding of a rigid diol with tetrabutylammonium chloride in acetonitrile-d3. A favorable carbon-oxygen dipole and an intramolecular C-HOH hydrogen bond in an equatorially substituted ether is found to be energetically more important than a stabilizing hydrogen bond in the corresponding axially oriented alcohol. IR spectroscopy is also used to probe the structures of the bound complexes and several binding motifs are identified.
Sigma-Aldrich
2,3-Dichloro-5,6-dicyano-p-benzoquinone, 98%
Sigma-Aldrich
Dess-Martin periodinane, 97%
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer