Merck
  • M2b macrophage elimination and improved resistance of mice with chronic alcohol consumption to opportunistic infections.
M2b macrophage elimination and improved resistance of mice with chronic alcohol consumption to opportunistic infections.
The American journal of pathology (2014-12-09)
Hideko Ohama, Akira Asai, Ichiaki Ito, Sumihiro Suzuki, Makiko Kobayashi, Kazuhide Higuchi, Fujio Suzuki
ABSTRACT
Alcohol abuse was found to predispose persons to opportunistic infections. In this study, we tried to improve the host antibacterial resistance of chronic alcohol-consuming (CAC) mice to opportunistic infections. Bactericidal macrophages with functions to produce IL-12 and to express mRNAs for CXCL9 and inducible nitric oxide synthase (M1 macrophages) were characterized as the main effector cells in host antibacterial innate immunities against infections with opportunistic pathogens. However, CAC mice were found to be carriers of M2b macrophages [macrophages with functions to produce IL-10 and to express mRNAs for CD163, chemokine ligand (CCL)1, and LIGHT (homologous to lymphotoxin, exhibits inducible expression, competes with herpes simplex virus glycoprotein D for high-voltage electron microscopy on T cells)], which were inhibitory on macrophage conversion from resident macrophages to M1 macrophages. Under treatment with CCL1 antisense oligodeoxynucleotides, a specific inhibitor of M2b macrophages, CAC mouse macrophages reverted to resident macrophages, and M1 macrophages were induced by a bacterial antigen from macrophages of CAC mice that were previously treated with the oligodeoxynucleotides. Opportunistic infections (enterococcal translocation and Klebsiella pneumonia) in CAC mice were completely controlled by CCL1 antisense oligodeoxynucleotides. These results indicate that certain opportunistic infections in alcoholics are controllable through the modulation of M2b macrophages.
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Gadolinium(III) chloride solution, NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), Methanol-13C 0.1 % (99 atom % 13C), water 1 %, NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Gadolinium(III) chloride solution, NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), water 0.1 %, NMR tube size 6.5 mm × 8 in.
Sigma-Aldrich
Gadolinium(III) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Gadolinium(III) chloride solution, NMR reference standard, water 1 %, DSS 0.1 %, 0.1 mg/mL in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology