Merck
  • Characterization of an H9N2 avian influenza virus from a Fringilla montifringilla brambling in northern China.
Characterization of an H9N2 avian influenza virus from a Fringilla montifringilla brambling in northern China.
Virology (2015-01-09)
Jing Yuan, Lili Xu, Linlin Bao, Yanfeng Yao, Wei Deng, Fengdi Li, Qi Lv, Songzhi Gu, Qiang Wei, Chuan Qin
ABSTRACT
Avian H9N2 influenza viruses circulating in domestic poultry populations are occasionally transmitted to humans. We report the genomic characterization of an H9N2 avian influenza virus (A/Brambling/Beijing/16/2012) first isolated from a healthy Fringilla montifringilla brambling in northern China in 2012. Phylogenetic analyses revealed that this H9N2 virus belongs to the BJ/94-like sublineage. This virus had a low pathogenicity for chickens and was able to replicate at a low level in mouse lung tissue. Transmission studies in ferrets showed that this H9N2 strain shed high levels of the virus in nasal and throat swabs. In vitro receptor binding assays, the virus bound only to α-2,6 linkage receptors and not to the avian-type α-2,3 linkage receptors, suggesting that H9N2 influenza viruses present potential public health risks. Therefore, attention should be paid to H9N2 influenza viruses and the close surveillance of H9N2 viruses in poultry.
Product Description
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
N-Acetylneuraminic acid, ≥95% anhydrous basis, synthetic
Sigma-Aldrich
N-Acetylneuraminic acid, ≥98% (HPLC), from Escherichia coli
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Cytidine-5′-monophospho-N-acetylneuraminic acid sodium salt, ≥85% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
USP
Galactose, United States Pharmacopeia (USP) Reference Standard
Galactose, European Pharmacopoeia (EP) Reference Standard
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material