Merck
  • Clove extract inhibits tumor growth and promotes cell cycle arrest and apoptosis.
Clove extract inhibits tumor growth and promotes cell cycle arrest and apoptosis.
Oncology research (2014-05-24)
Haizhou Liu, John C Schmitz, Jianteng Wei, Shousong Cao, Jan H Beumer, Sandra Strychor, Linyou Cheng, Ming Liu, Cuicui Wang, Ning Wu, Xiangzhong Zhao, Yuyan Zhang, Joshua Liao, Edward Chu, Xiukun Lin
ABSTRACT
Cloves (Syzygium aromaticum) have been used as a traditional Chinese medicinal herb for thousands of years. Cloves possess antiseptic, antibacterial, antifungal, and antiviral properties, but their potential anticancer activity remains unknown. In this study, we investigated the in vitro and in vivo antitumor effects and biological mechanisms of ethyl acetate extract of cloves (EAEC) and the potential bioactive components responsible for its antitumor activity. The effects of EAEC on cell growth, cell cycle distribution, and apoptosis were investigated using human cancer cell lines. The molecular changes associated with the effects of EAEC were analyzed by Western blot and (qRT)-PCR analysis. The in vivo effect of EAEC and its bioactive component was investigated using the HT-29 tumor xenograft model. We identified oleanolic acid (OA) as one of the components of EAEC responsible for its antitumor activity. Both EAEC and OA display cytotoxicity against several human cancer cell lines. Interestingly, EAEC was superior to OA and the chemotherapeutic agent 5-fluorouracil at suppressing growth of colon tumor xenografts. EAEC promoted G0/G1 cell cycle arrest and induced apoptosis in a dose-dependent manner. Treatment with EAEC and OA selectively increased protein expression of p21(WAF1/Cip1) and γ-H2AX and downregulated expression of cell cycle-regulated proteins. Moreover, many of these changes were at the mRNA level, suggesting transcriptional regulation by EAEC treatment. Our results demonstrate that clove extract may represent a novel therapeutic herb for the treatment of colorectal cancer, and OA appears to be one of the bioactive components.
Sigma-Aldrich
Anti-Survivin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Supelco
Eugenol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Eugenol, European Pharmacopoeia (EP) Reference Standard
Supelco
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material