Merck
  • Evaluation of sorbent materials for the sampling and analysis of phosphine, sulfuryl fluoride and methyl bromide in air.
Evaluation of sorbent materials for the sampling and analysis of phosphine, sulfuryl fluoride and methyl bromide in air.
Journal of chromatography. A (2014-12-17)
R Magnusson, L Rittfeldt, C Åstot
ABSTRACT
Phosphine (PH3), sulfuryl fluoride (SO2F2) and methyl bromide (CH3Br) are highly toxic chemical substances commonly used for fumigation, i.e., pest control with gaseous pesticides. Residues of fumigation agents constitute a health risk for workers affected, and therefore accurate methods for air sampling and analysis are needed. In this study, three commercial adsorbent tubes; Carbosieve SIII™, Air Toxics™ and Tenax TA™, were evaluated for sampling these highly volatile chemicals in air and their subsequent analysis by thermal desorption-gas chromatography-mass spectrometry (TD-GC-MS). The breakthrough volume (BTV) of each fumigant was experimentally determined on the different adsorbents at concentrations at or above their permissible exposure limits, using a method based on frontal chromatography of generated fumigant atmospheres. Carbosieve SIII™, a molecular sieve possessing a very high specific area, proved to be a better adsorbent than both Air Toxics™ and Tenax TA™, resulting in at least a 4-fold increase of the BTV50%. BTV50% for Carbosieve SIII™ at 20°C was measured as 4.7L/g, 5.5L/g and 126L/g for phosphine, sulfuryl fluoride and methyl bromide, respectively, implying safe sampling volumes of 1.9L, 2.2L and 50L, respectively, for a commercial tube packed with 800mg Carbosieve SIII™. The temperature dependence of BTV was strong for Carbosieve SIII™, showing a reduction of 3-5%/°C in breakthrough volume within the range -20 to 40°C. Furthermore, although Carbosieve SIII™ reportedly has a higher affinity for water than most other adsorbents, relative humidity had only a moderate influence on the retention capacity of phosphine. Overall, the applicability of Carbosieve SIII™ adsorbent sampling in combination with TD-GC-MS analysis was demonstrated for highly volatile fumigants.
Product Description
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-III, 60-80 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-II, 60-80 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve G, 60-80 mesh, bottle of 5 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve G, 80-100 mesh, bottle of 5 g
Sigma-Aldrich
Nitrogen, ≥99.998%
Supelco
Carbosieve SIII, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-II, 80-100 mesh, bottle of 10 g