Merck
  • Hyperspectral imaging in quality control of inkjet printed personalised dosage forms.
Hyperspectral imaging in quality control of inkjet printed personalised dosage forms.
International journal of pharmaceutics (2014-12-21)
Hossein Vakili, Ruzica Kolakovic, Natalja Genina, Mathieu Marmion, Harri Salo, Petri Ihalainen, Jouko Peltonen, Niklas Sandler
ABSTRACT
The aim of the study was to investigate applicability of near infra-red (NIR) hyperspectral imaging technique in quality control of printed personalised dosage forms. Inkjet printing technology was utilized to fabricate escalating doses of an active pharmaceutical ingredient (API). A solution containing anhydrous theophylline as the model drug was developed as a printable formulation. Single units solid dosage forms (SDFs) were prepared by jetting the solution onto 1 cm × 1 cm areas on carrier substrate with multiple printing passes. It was found that the number of printing passes was in excellent correlation (R(2)=0.9994) with the amount of the dispensed drug (μg cm(-2)) based on the UV calibration plot. The API dose escalation was approximately 7.5 μg cm(-2) for each printing pass concluding that inkjet printing technology can optimally provide solutions to accurate deposition of active substances with a potential for personalized dosing. Principal component analysis (PCA) was carried out in order to visualize the trends in the hyperspectral data. Subsequently, a quantitative partial least squares (PLS) regression model was created. NIR hyperspectral imaging proved (R(2)=0.9767) to be a reliable, rapid and non-destructive method to optimize quality control of these planar printed dosage forms.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Supelco
Theophylline, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Theophylline, United States Pharmacopeia (USP) Reference Standard
Theophylline, European Pharmacopoeia (EP) Reference Standard
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycerol, analytical standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%