Merck
  • Urinary kidney injury molecule 1 (KIM-1) and interleukin 18 (IL-18) as risk markers for heart failure in older adults: the Health, Aging, and Body Composition (Health ABC) Study.
Urinary kidney injury molecule 1 (KIM-1) and interleukin 18 (IL-18) as risk markers for heart failure in older adults: the Health, Aging, and Body Composition (Health ABC) Study.
American journal of kidney diseases : the official journal of the National Kidney Foundation (2014-03-25)
Todd H Driver, Ronit Katz, Joachim H Ix, Jared W Magnani, Carmen A Peralta, Chirag R Parikh, Linda Fried, Anne B Newman, Stephen B Kritchevsky, Mark J Sarnak, Michael G Shlipak
ABSTRACT
Kidney damage and reduced kidney function are potent risk factors for heart failure, but existing studies are limited to assessing albuminuria or estimated glomerular filtration rate (eGFR). We evaluated the associations of levels of urinary biomarkers of kidney tubular injury (interleukin 18 [IL-18] and kidney injury molecule 1 [KIM-1]) with future risk of heart failure. Retrospective cohort study. 2,917 participants without heart failure in the Health, Aging, and Body Composition (Health ABC) cohort. Ratios of urine KIM-1, IL-18, and albumin to creatinine (KIM-1:Cr, IL-18:Cr, and ACR, respectively). Incident heart failure over a median follow-up of 12 years. Median values of each marker at baseline were 812 (IQR, 497-1,235)pg/mg for KIM-1:Cr, 31 (IQR, 19-56)pg/mg for IL-18:Cr, and 8 (IQR, 5-19) mg/g for ACR. 596 persons developed heart failure during follow-up. The top quartile of KIM-1:Cr was associated with risk of incident heart failure after adjustment for baseline eGFR, heart failure risk factors, and ACR (HR, 1.32; 95% CI, 1.02-1.70) in adjusted multivariate proportional hazards models. The top quartile of IL-18:Cr also was associated with heart failure in a model adjusted for risk factors and eGFR (HR, 1.35; 95% CI, 1.05-1.73), but was attenuated by adjustment for ACR (HR, 1.15; 95% CI, 0.89-1.48). The top quartile of ACR had a stronger adjusted association with heart failure (HR, 1.96; 95% CI, 1.53-2.51). Generalizability to other populations is uncertain. Higher urine KIM-1 concentrations were associated independently with incident heart failure risk, although the associations of higher ACR were of stronger magnitude.
Product Description
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Human HAVCR1 / Hepatitis A Virus Cellular Receptor 1 ELISA Kit
Sigma-Aldrich
Mouse Havcr1 / Hepatitis A Virus Cellular Receptor 1 Homolog ELISA Kit
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Rat Havcr1 / Hepatitis A Virus Cellular Receptor 1 Homolog ELISA Kit
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications