Merck
  • Cell cycle-related kinase mediates viral-host signalling to promote hepatitis B virus-associated hepatocarcinogenesis.
Cell cycle-related kinase mediates viral-host signalling to promote hepatitis B virus-associated hepatocarcinogenesis.
Gut (2014-01-21)
Zhuo Yu, Yue-Qiu Gao, Hai Feng, Ying-Ying Lee, May S Li, Yuan Tian, Minnie Y Y Go, Dae-Yeul Yu, Yue-Sun Cheung, Paul B S Lai, Jun Yu, Vincent W S Wong, Joseph J Y Sung, Henry L Y Chan, Alfred S L Cheng
ABSTRACT
Androgen receptor (AR) signalling contributes to male predominance in hepatocellular carcinoma (HCC), which is more pronounced in HBV-endemic areas. Cell cycle-related kinase (CCRK) is essential for AR-induced hepatocarcinogenesis but its molecular function in HBV-associated HCC remains obscure. To determine the molecular function of CCRK in HBV-associated HCC. Transcriptional regulation was assessed by chromatin immunoprecipitation, promoter mutation and luciferase reporter assays. Hepatocellular proliferation and tumourigenesis were examined by colony formation, soft agar assays and using HBV X protein (HBx) transgenic mice with low-dose exposure to diethylnitrosamine. Protein expressions were examined in clinical samples and correlated with patient survival by log-rank Mantel-Cox test. Overexpression of CCRK, but not its kinase-defective mutant, activated β-catenin/T cell factor signalling through phosphorylation of glycogen synthase kinase-3β (GSK-3β) at Ser9, led to upregulation of AR transcriptional activity and, subsequently, expression of HBx. The viral transactivator in turn induced CCRK expression through enhanced AR signalling, thus forming a positive regulatory loop. RNA interference silencing of CCRK, which suppressed the CCRK/GSK-3β/β-catenin/AR regulatory loop, significantly suppressed HBx-induced hepatocellular proliferation (p=0.001) and transformation (p<0.001) and remarkably reduced >80% diethylnitrosamine-mediated hepatocarcinogenesis in HBx transgenic mice. Finally, patients with HBV-associated HCC with concordant overexpression of CCRK, GSK-3β phosphorylation at Ser9, active dephosphorylated β-catenin and AR phosphorylation at Ser81 had poorer overall (HR=31.26, p<0.0001) and disease-free (HR=3.60, p<0.01) survival rates. Our findings highlight the critical role of CCRK in a self-reinforcing circuitry that regulates HBV-associated hepatocarcinogenesis. Further characterisation of this intricate viral-host signalling may provide new prognostic biomarkers and therapeutic targets for HCC treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ccrk
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Supelco
N-Nitrosodiethylamine, analytical standard
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
N-Nitrosodiethylamine, ISOPAC®
Sigma-Aldrich
N-Nitrosodiethylamine, liquid
Sigma-Aldrich
N-Nitrosodiethylamine, ≥99.0% (GC)
Sigma-Aldrich
MISSION® esiRNA, targeting human CDK20