Merck
  • Implementing visible 473 nm photodissociation in a Q-Exactive mass spectrometer: towards specific detection of cysteine-containing peptides.
Implementing visible 473 nm photodissociation in a Q-Exactive mass spectrometer: towards specific detection of cysteine-containing peptides.
The Analyst (2014-09-10)
Marion Girod, Jordane Biarc, Quentin Enjalbert, Arnaud Salvador, Rodolphe Antoine, Philippe Dugourd, Jérôme Lemoine
ABSTRACT
Improvement of the fragmentation specificity may streamline data processing of bottom-up proteomic experiments by drastically reducing either the amount of MS/MS data to process in the discovery phase or the detection of interfering signals in targeted quantification. Photodissociation at appropriate wavelengths is a promising alternative technique to the non-discriminating conventional activation mode by collision. Here, we describe the implementation of visible LID at 473 nm in a Q-Exactive-Orbitrap mass spectrometer for the specific detection of cysteine-containing peptides tagged with a Dabcyl group. HCD cell DC offset and irradiation time were optimized to obtain high fragmentation yield and spectra free of contaminating CID product ions, while keeping the irradiation time scale compatible with chromatographic separation. With this optimized experimental set-up, the selective detection of cysteine-containing peptides in a whole tryptic hydrolysate of three combined proteins is demonstrated by comparing all ion fragmentation (AIF) spectra recorded online with and without laser irradiation.
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
SAFC
Iodoacetamide
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%