Merck
  • The role of oxytocin in mothers' theory of mind and interactive behavior during the perinatal period.
The role of oxytocin in mothers' theory of mind and interactive behavior during the perinatal period.
Psychoneuroendocrinology (2014-07-06)
Anna L MacKinnon, Ian Gold, Nancy Feeley, Barbara Hayton, C Sue Carter, Phyllis Zelkowitz
ABSTRACT
The present longitudinal study examined the relations between plasma oxytocin, theory of mind, and maternal interactive behavior during the perinatal period. A community sample of women was assessed at 12-14 weeks gestation, 32-34 weeks gestation, and 7-9 weeks postpartum. Oxytocin during late pregnancy was significantly positively correlated with a measure of theory of mind, and predicted theory of mind ability after controlling for parity, maternal education, prenatal psychosocial risk, and general anxiety, measured during the first trimester. Theory of mind was associated with less remote and less depressive maternal interactive behavior. Oxytocin, across all time points, was not directly related to maternal interactive behavior. However, there was a significant indirect effect of oxytocin during late pregnancy on depressive maternal behavior via theory of mind ability. These preliminary findings suggest that changes in the oxytocinergic system during the perinatal period may contribute to the awareness of social cues, which in turn plays a role in maternal interactive behavior.
Sigma-Aldrich
Oxytocin
Sigma-Aldrich
Oxytocin, lyophilized powder, ~15 IU/mg solid (Prepared from synthetic oxytocin)
USP
Oxytocin, United States Pharmacopeia (USP) Reference Standard
Oxytocin, European Pharmacopoeia (EP) Reference Standard