Merck
Azide-tagged sphingolipids: new tools for metabolic flux analysis.
Chembiochem : a European journal of chemical biology (2015-02-14)
María Garrido, José Luis Abad, Gemma Fabriàs, Josefina Casas, Antonio Delgado
ABSTRACT
Several diseases involve alterations in sphingolipid metabolism, so the development of tools for the analysis of sphingolipid metabolic fluxes is of interest. In this work, ω-azidosphingolipids 1-3 have been synthesized and tested as tracers in live cells. The synthesis starts from (S)-Garner's aldehyde and uses bromide or tosyloxy precursors for the introduction of the azido group into the sphingoid base. Studies in HGC-27 cells showed that probes 1-3 compete with the natural metabolites and are incorporated into sphingolipid pathways without affecting cell viability. The reactivity and bioorthogonality of the terminal azido group have been exploited by means of click reactions with different azadibenzocyclooctyne tags. This allows the mass spectrometric characterization of azidosphingolipidomes in pooled samples from different cell populations after independent treatments, providing proof of concept of the applicability of this technology in sphingolipid metabolic flux analysis.
Product Description
Supelco
Methanol, analytical standard
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Rhodium, powder, 99.95% trace metals basis
Sigma-Aldrich
Rhodium on alumina, extent of labeling: 5 wt. % loading, matrix alumina support, powder
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.0005mm, permanent polyester support, 99.85%
Rhodium, microfoil, disks, 10mm, thinness 0.1μm, specific density 124μg/cm2, permanent mylar 3.5μm support, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.85%
Rhodium, foil, light tested, 15x15mm, thickness 0.05mm, as rolled, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.00075mm, permanent polyester support, 99.85%
Rhodium, foil, not light tested, 25x25mm, thickness 0.006mm, as rolled, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.012mm, as rolled, 99.9%
Rhodium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Rhodium, microfoil, disks, 10mm, thinness 0.50μm, specific density 620μg/cm2, permanent mylar 3.5μm support, 99.99%
Rhodium, foil, not light tested, 50x50mm, thickness 0.00025mm, permanent polyester support, 99.9%
Rhodium, microfoil, disks, 25mm, thinness 0.1μm, specific density 124μg/cm2, permanent mylar 3.5μm support, 99.9%
Rhodium, wire reel, 0.05m, diameter 0.12mm, as drawn, 99.9%
Rhodium, wire reel, 0.05m, diameter 0.25mm, hard, 99.9%
Rhodium, foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 99.9%