Merck
  • Metabolic imaging of pancreatic ductal adenocarcinoma detects altered choline metabolism.
Metabolic imaging of pancreatic ductal adenocarcinoma detects altered choline metabolism.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-11-06)
Marie-France Penet, Tariq Shah, Santosh Bharti, Balaji Krishnamachary, Dmitri Artemov, Yelena Mironchik, Flonné Wildes, Anirban Maitra, Zaver M Bhujwalla
ABSTRACT
Pancreatic ductal adenocarcinoma (PDAC) is an aggressive and lethal disease that develops relatively symptom-free and is therefore advanced at the time of diagnosis. The absence of early symptoms and effective treatments has created a critical need for identifying and developing new noninvasive biomarkers and therapeutic targets. We investigated the metabolism of a panel of PDAC cell lines in culture and noninvasively in vivo with (1)H magnetic resonance spectroscopic imaging (MRSI) to identify noninvasive biomarkers and uncover potential metabolic targets. We observed elevated choline-containing compounds in the PDAC cell lines and tumors. These elevated choline-containing compounds were easily detected by increased total choline (tCho) in vivo, in spectroscopic images obtained from tumors. Principal component analysis of the spectral data identified additional differences in metabolites between immortalized human pancreatic cells and neoplastic PDAC cells. Molecular characterization revealed overexpression of choline kinase (Chk)-α, choline transporter 1 (CHT1), and choline transporter-like protein 1 (CTL1) in the PDAC cell lines and tumors. Collectively, these data identify new metabolic characteristics of PDAC and reveal potential metabolic targets. Total choline detected with (1)H MRSI may provide an intrinsic, imaging probe-independent biomarker to complement existing techniques in detecting PDAC. The expression of Chk-α, CHT1, and CTL1 may provide additional molecular markers in aspirated cytological samples.
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Sodium Fluoride Solution
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Anti-GAPDH antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)