Merck
  • Intercellular bridges are essential for human parthenogenetic cell survival.
Intercellular bridges are essential for human parthenogenetic cell survival.
Mechanisms of development (2015-02-24)
Georgia Pennarossa, Sara Maffei, Gianluca Tettamanti, Terenzio Congiu, Magda deEguileor, Fulvio Gandolfi, Tiziana A L Brevini
ABSTRACT
Parthenogenetic cells, obtained from in vitro activated mammalian oocytes, display multipolar spindles, chromosome malsegregation and a high incidence of aneuploidy, probably due to the lack of paternal contribution. Despite this, parthenogenetic cells do not show high rates of apoptosis and are able to proliferate in a way comparable to their biparental counterpart. We hypothesize that a series of adaptive mechanisms are present in parthenogenetic cells, allowing a continuous proliferation and ordinate cell differentiation both in vitro and in vivo. Here we identify the presence of intercellular bridges that contribute to the establishment of a wide communication network among human parthenogenetic cells, providing a mutual exchange of missing products. Silencing of two molecules essential for intercellular bridge formation and maintenance demonstrates the key function played by these cytoplasmic passageways that ensure normal cell functions and survival, alleviating the unbalance in cellular component composition.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology