- CO/C-H as an acylating reagent: a palladium-catalyzed aerobic oxidative carbonylative esterification of alcohols.
CO/C-H as an acylating reagent: a palladium-catalyzed aerobic oxidative carbonylative esterification of alcohols.
Angewandte Chemie (International ed. in English) (2014-04-18)
Lu Wang, Yanxia Wang, Chao Liu, Aiwen Lei
PMID24740697
ABSTRACT
A palladium-catalyzed oxidative carbonylative esterification of a variety of functionalized alcohols under base- and ligand-free conditions has been demonstrated. A CO/olefin combination was utilized as the acylating reagent with O2 as a benign oxidant. Notably, the scope of the substrate alcohols has been greatly broadened.