Merck
  • The newly synthesized anticancer drug HUHS1015 is useful for treatment of human gastric cancer.
The newly synthesized anticancer drug HUHS1015 is useful for treatment of human gastric cancer.
Cancer chemotherapy and pharmacology (2015-01-09)
Yoshiko Kaku, Ayako Tsuchiya, Takeshi Kanno, Shuhei Nakao, Tadashi Shimizu, Akito Tanaka, Tomoyuki Nishizaki
ABSTRACT
Naftopidil is clinically for treatment of benign prostate hyperplasia, and emerging evidence has pointed to its anticancer effect. To obtain the anticancer drug with the potential greater than that of naftopidil, we have newly synthesized the naftopidil analogue HUHS1015. The present study investigated the mechanism underlying HUHS1015-induced apoptosis of human gastric cancer cells and assessed the possibility for clinical use as an innovative anticancer drug. HUHS1015 reduced cell viability for MKN28 human well-differentiated gastric adenocarcinoma cell line and MKN45 human poorly differentiated gastric adenocarcinoma cell line in a concentration (0.3-100 μM)-dependent manner more effectively than cisplatin, a chemo-drug widely used. In the flow cytometry using propidium iodide (PI) and annexin V, HUHS1015 significantly increased the population of PI-positive and annexin V-negative cells, corresponding to primary necrosis and that of PI-positive and annexin V-positive cells, corresponding to late apoptosis/secondary necrosis, both in the two cell types. HUHS1015 significantly activated caspase-3, caspase-4, and caspase-8 in MKN45 cells, while no obvious caspase activation was found in MKN28 cells. HUHS1015 upregulated expression of the tumor necrosis factor α (TNFα) mRNA and protein in MKN45 cells, allowing activation of caspase-8 through TNF receptor and the effector caspase-3. HUHS1015 clearly inhibited tumor growth in mice inoculated with MKN45 cells, with the survival rate higher than that for the anticancer drugs cisplatin, paclitaxel, and irinotecan. The results of the present study show that HUHS1015 induces caspase-independent and caspase-dependent apoptosis of MKN28 and MKN45 human gastric cancer cells, respectively, and effectively suppresses MKN45 cell proliferation.
Product Description
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%