Merck
  • Elimination of damaged mitochondria through mitophagy reduces mitochondrial oxidative stress and increases tolerance to trichothecenes.
Elimination of damaged mitochondria through mitophagy reduces mitochondrial oxidative stress and increases tolerance to trichothecenes.
Proceedings of the National Academy of Sciences of the United States of America (2014-07-30)
Mohamed Anwar Bin-Umer, John E McLaughlin, Matthew S Butterly, Susan McCormick, Nilgun E Tumer
ABSTRACT
Trichothecene mycotoxins are natural contaminants of small grain cereals and are encountered in the environment, posing a worldwide threat to human and animal health. Their mechanism of toxicity is poorly understood, and little is known about cellular protection mechanisms against trichothecenes. We previously identified inhibition of mitochondrial protein synthesis as a novel mechanism for trichothecene-induced cell death. To identify cellular functions involved in trichothecene resistance, we screened the Saccharomyces cerevisiae deletion library for increased sensitivity to nonlethal concentrations of trichothecin (Tcin) and identified 121 strains exhibiting higher sensitivity than the parental strain. The largest group of sensitive strains had significantly higher reactive oxygen species (ROS) levels relative to the parental strain. A dose-dependent increase in ROS levels was observed in the parental strain treated with different trichothecenes, but not in a petite version of the parental strain or in the presence of a mitochondrial membrane uncoupler, indicating that mitochondria are the main site of ROS production due to toxin exposure. Cytotoxicity of trichothecenes was alleviated after treatment of the parental strain and highly sensitive mutants with antioxidants, suggesting that oxidative stress contributes to trichothecene sensitivity. Cotreatment with rapamycin and trichothecenes reduced ROS levels and cytotoxicity in the parental strain relative to the trichothecene treatment alone, but not in mitophagy deficient mutants, suggesting that elimination of trichothecene-damaged mitochondria by mitophagy improves cell survival. These results reveal that increased mitophagy is a cellular protection mechanism against trichothecene-induced mitochondrial oxidative stress and a potential target for trichothecene resistance.
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
Supelco
(±)-α-Tocopherol, analytical standard
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Alpha Tocopherol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
α-Tocopherol, ≥95.5%
α-Tocopherol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard