Merck
Reaction of nucleosome DNA with dimethyl sulfate.
Proceedings of the National Academy of Sciences of the United States of America (1979-05-01)
J D McGhee, G Felsenfeld
PMID287052
ABSTRACT
We have measured the effect of the histones in the nucleosome core particle on methylation of purines in nucleosome DNA by dimethyl sulfate. By using 32P terminally labeled nucleosome cores, we have examined the pattern of strand cleavage at methylated sites in the nucleosome DNA and compared it to the pattern observed in histone-free DNA. We are unable to detect any significant difference between the reactivity of N7 of guanines in nucleosome DNA and of that in naked DNA, with the exception of a single site of enhanced reactivity at approximately nucleotide 62 from the 5' end of the nucleosome. Contrary to our expectation, there is no detectable periodic modulation of reactivity corresponding to the twist of the DNA on the nucleosome surface. We are able to place a low upper limit on the extent to which the histones of the nucleosome can protect N7 of guanine in the large groove. With somewhat less precision, we also conclude that the N3 of adenine in the small groove is largely unprotected. These results indicate that in nucleosome DNA the bases are nearly as accessible to solvent as they are in DNA free of protein.
Sigma-Aldrich
Dimethyl sulfate, puriss. p.a., ≥99.0% (GC)