Merck
  • Programmable periodicity of quantum dot arrays with DNA origami nanotubes.
Programmable periodicity of quantum dot arrays with DNA origami nanotubes.
Nano letters (2010-08-05)
Hieu Bui, Craig Onodera, Carson Kidwell, YerPeng Tan, Elton Graugnard, Wan Kuang, Jeunghoon Lee, William B Knowlton, Bernard Yurke, William L Hughes
ABSTRACT
To fabricate quantum dot arrays with programmable periodicity, functionalized DNA origami nanotubes were developed. Selected DNA staple strands were biotin-labeled to form periodic binding sites for streptavidin-conjugated quantum dots. Successful formation of arrays with periods of 43 and 71 nm demonstrates precise, programmable, large-scale nanoparticle patterning; however, limitations in array periodicity were also observed. Statistical analysis of AFM images revealed evidence for steric hindrance or site bridging that limited the minimum array periodicity.
Sigma-Aldrich
Magnesium acetate tetrahydrate, ACS reagent, ≥98%