Merck
  • Comparison of mineral and soluble iron Fenton's catalysts for the treatment of trichloroethylene.
Comparison of mineral and soluble iron Fenton's catalysts for the treatment of trichloroethylene.
Water research (2001-03-10)
A L Teel, C R Warberg, D A Atkinson, R J Watts
ABSTRACT
Contaminant degradation, stoichiometry, and role of hydroxyl radicals (OH*) in four Fenton's systems were investigated using trichloroethylene (TCE) as a model contaminant. A standard Fenton's system, a modified soluble iron system with a pulse input of hydrogen peroxide, and two modified mineral-catalyzed systems (pH 3 and 7) were studied. In the standard Fenton's system, which had the most efficient reaction stoichiometry, 78% of the TCE was degraded; however, chloride analysis indicated that no more than two of the three chlorines were displaced per TCE molecule degraded. Although the modified soluble iron system was characterized by 91% TCE degradation, chloride analysis also indicated that no more than two of the chlorines were lost from the TCE. In the goethite system of pH 3, > 99% of the TCE was degraded. Near-complete release of chloride suggested that the TCE may have been mineralized. Only 22% degradation of TCE was achieved in the pH 7 goethite system. and there was minimal release of chloride. The mineral-catalyzed reactions exhibited the least efficient reaction stoichiometry of the four systems. Experiments using hydroxyl radical scavengers showed that the standard Fenton's system degraded TCE entirely by hydroxyl radical mechanisms, while approximately 10-15% of the degradation achieved in the modified soluble iron and goethite-catalyzed systems at pH 3 was mediated by non-hydroxyl radical mechanisms. In the goethite system at pH 7, only non-hydroxyl radical mechanisms were found. The goethite-catalyzed system at pH 3 effectively degraded the parent compound and may have the potential to mineralize contaminants when used for in situ soil and groundwater remediation and ex situ waste stream treatment in packed-bed reactors.
Sigma-Aldrich
Trichloroethylene, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%