Merck
  • Design and optimization of hybrid of 2,4-diaminopyrimidine and arylthiazole scaffold as anticancer cell proliferation and migration agents.
Design and optimization of hybrid of 2,4-diaminopyrimidine and arylthiazole scaffold as anticancer cell proliferation and migration agents.
European journal of medicinal chemistry (2015-04-23)
Wenbo Zhou, Anling Huang, Yong Zhang, Qingxiang Lin, Weikai Guo, Zihua You, Zhengfang Yi, Mingyao Liu, Yihua Chen
ABSTRACT
Therapeutics of metastatic or triple-negative breast cancer are still challenging in clinical. Herein we demonstrated the design and optimization of a series of hybrid of 2,4-diaminopyrimidine and arylthiazole derivatives for their anti-proliferative properties against two breast cancer cell lines (MCF-7 as human breast cancer and MDA-MB-231 as triple-negative breast cancer). More importantly, some of those compounds with potent antiproliferative activities also indicated excellent inhibitory activities against MDA-MB-231 cell migration. These results suggested that the new series of hybridation of aryl-thiazoles and aminopyrimidines could be identified and developed as novel highly potential anticancer agents against the triple-negative breast cancer as well as metastatic one in the future.
Sigma-Aldrich
2,4-Diaminopyrimidine, 98%