Merck
  • Influence of double-strand break repair on radiation therapy-induced acute skin reactions in breast cancer patients.
Influence of double-strand break repair on radiation therapy-induced acute skin reactions in breast cancer patients.
International journal of radiation oncology, biology, physics (2014-01-15)
Kamalesh Dattaram Mumbrekar, Donald Jerard Fernandes, Hassan Venkatesh Goutham, Krishna Sharan, Bejadi Manjunath Vadhiraja, Kapaettu Satyamoorthy, Satish Rao Bola Sadashiva
ABSTRACT
Curative radiation therapy (RT)-induced toxicity poses strong limitations for efficient RT and worsens the quality of life. The parameter that explains when and to what extent normal tissue toxicity in RT evolves would be of clinical relevance because of its predictive value and may provide an opportunity for personalized treatment approach. DNA double-strand breaks and repair were analyzed by microscopic γ-H2AX foci analysis in peripheral lymphocytes from 38 healthy donors and 80 breast cancer patients before RT, a 2 Gy challenge dose of x-ray exposed in vitro. The actual damage (AD) at 0.25, 3, and 6 hours and percentage residual damage (PRD) at 3 and 6 hours were used as parameters to measure cellular radiosensitivity and correlated with RT-induced acute skin reactions in patients stratified as non-overresponders (NOR) (Radiation Therapy Oncology Group [RTOG] grade <2) and overresponders (OR) (RTOG grade ≥2). The results indicated that the basal and induced (at 0.25 and 3 hours) γ-H2AX foci numbers were nonsignificant (P>.05) between healthy control donors and the NOR and OR groups, whereas it was significant between ORs and healthy donors at 6 hours (P<.001). There was a significantly higher PRD in OR versus NOR (P<.05), OR versus healthy donors (P<.001) and NOR versus healthy donors (P<.01), supported further by the trend analysis (r=.2392; P=.0326 at 6 hours). Our findings strongly suggest that the measurement of PRD by performing γ-H2AX foci analysis has the potential to be developed into a clinically useful predictive assay.
Product Description
Ficoll® Paque Plus, Cytiva 17-1440-03, pack of 6 × 500 mL