Merck
  • BIN1 membrane curvature sensing and generation show autoinhibition regulated by downstream ligands and PI(4,5)P2.
BIN1 membrane curvature sensing and generation show autoinhibition regulated by downstream ligands and PI(4,5)P2.
Biochemistry (2014-10-29)
Tingting Wu, Tobias Baumgart
ABSTRACT
In striated muscles, invaginations from the plasma membrane, termed transverse tubules (T-tubule), function in the excitation-contraction coupling machinery. BIN1 (isoform8) plays a critical role in the biogenesis of T-tubules. BIN1 contains an N-terminal BAR domain to sense and induce membrane curvature, an isoform8-specific polybasic motif (exon10) as the phosphoinositide binding module and a C-terminal Src homology 3 (SH3) domain for the recruitment of downstream proteins such as dynamin 2. Previous studies of N-BAR domains focused on elucidating mechanisms of membrane curvature sensing and generation (MC-S&G). Less is known about how MC-S&G is regulated. We found that the SH3 domain binds to the exon10 motif more strongly compared to the proline-rich domain (PRD) of dynamin 2. Furthermore, we found that the MC-S&G ability of full-length BIN1 is inhibited on membranes lacking PI(4,5)P2. Addition of PI(4,5)P2 in the membrane activates BIN1 to sense and induce membrane curvature. Co-presence of the SH3 domain and exon10 motif leads to the strongest phosphoinositide-mediated control of BIN1 function. Addition of SH3 domain ligand (such as PRD peptides), as well as addition of the water-soluble PI(4,5)P2 analogue, can both enhance the MC-S&G ability of BIN1 on membranes without PI(4,5)P2, indicating that the key to activate BIN1 is to disrupt the exon10-SH3 interaction. The nonsense mutation K436X, found in centronuclear myopathy (CNM) patients, abolishes SH3 domain binding with either exon10 or the PRD motif, resulting in increased membrane deformation capacity. Our results suggest an autoinhibition model for BIN1 that involves a synergistic regulation by membrane composition and protein-protein interactions.
Product Description
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, ≥97.0% (TLC)
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, ≥99.0% (10 mg phospholipid per ml CHCl3, TLC)
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications