Merck
  • Prevalence and characteristics of pancreatic cancer in families with BRCA1 and BRCA2 mutations.
Prevalence and characteristics of pancreatic cancer in families with BRCA1 and BRCA2 mutations.
Familial cancer (2008-10-16)
Daniel H Kim, Beth Crawford, John Ziegler, Mary S Beattie
ABSTRACT
A growing body of research describes cancers other than breast and ovarian in families with BRCA1/2 mutations, but the prevalence and characteristics of pancreatic cancer in these families has not been well described. This study was designed to: (1) estimate the prevalence of pancreatic cancer in BRCA1/2 positive families; (2) ascertain age of onset and gender distribution of pancreatic cancer in this cohort; and (3) compare age and gender characteristics of pancreatic cancer in BRCA1/2 positive families with those of the general population. Within the UCSF Cancer Risk Program cohort, 24/219 (11.0%) BRCA1 and 17/156 (10.9%) BRCA2 families had at least 1 individual with pancreatic cancer. In the 24 BRCA1 families, median age of diagnosis was 59 (range 45-80) in males, and 68 (range 38-87) in females (male:female ratio = 2.00). In the 17 BRCA2 families, median age of diagnosis was 67 (range 39-78) in males and 59 (range 46-81) in females (male:female ratio = 1.11). The SEER database, which describes cancer characteristics in a representative sample of the US population, reports a median age of 70 in males and 74 in females (male:female ratio = 0.96) over the same time period. Additionally, mean ages of diagnosis of pancreatic cancer in BRCA1/2 families differ significantly from the SEER mean (P = 0.0014 for BRCA1 and P = 0.011 for BRCA2 by unpaired t-test). Our findings suggest that families with early onset pancreatic cancer and features of hereditary breast and ovarian cancer should be considered for BRCA1/2 testing.