Merck
  • Chiral Brønsted acid catalyzed enantioselective synthesis of anti-homopropargyl alcohols via kinetic resolution-aldehyde allenylboration using racemic allenylboronates.
Chiral Brønsted acid catalyzed enantioselective synthesis of anti-homopropargyl alcohols via kinetic resolution-aldehyde allenylboration using racemic allenylboronates.
Organic letters (2013-03-15)
Andy S Tsai, Ming Chen, William R Roush
ABSTRACT
A chiral phosphoric acid catalyzed kinetic resolution/allenylboration of racemic allenylboronates with aldehydes is described. Allenylboration of aldehydes with 2.8 equiv of allenylboronate (±)-1 in the presence of 10 mol % of catalyst (R)-2 provided anti-homopropargyl alcohols 3 in 83-95% yield with 9:1 to 20:1 diastereoselectivity and 73-95% ee. The catalyst enables the kinetic resolution of the racemic allenylboronate (±)-1 to set the methyl stereocenter and biases the facial attack of the aldehyde to set the stereochemistry of the hydroxyl group in 3.
Sigma-Aldrich
Hydrocinnamaldehyde, technical grade, 90%
Pricing and availability is not currently available.