Merck
  • p8 Deficiency causes siderosis in spleens and lymphocyte apoptosis in acute pancreatitis.
p8 Deficiency causes siderosis in spleens and lymphocyte apoptosis in acute pancreatitis.
Pancreas (2014-08-08)
Sebastian Weis, Tilmann Cornelius Schlaich, Faramarz Dehghani, Tânia Carvalho, Ines Sommerer, Stephan Fricke, Franka Kahlenberg, Joachim Mössner, Albrecht Hoffmeister
ABSTRACT
The gene p8 was initially described in pancreatic tissue during acute experimental pancreatitis, a disease that is characterized by a systemic immune response. Although early reports suggested that p8 affects leukocyte migration during acute pancreatitis (AP), no studies revealing its immune-modulatory effects have been performed. We investigated the composition of the cellular immune system in naive p8 knockout (p8(−/−)) mice and compared with matched wild-type mice during pancreatitis. In young mice, there were no relevant differences in the composition of peripheral and splenic CD3(+), CD3(+)CD4(+), CD3(+)CD8(+), CD11b(+)Gr-1(-), and Gr-1 cells. In mature p8(−/−) mice, increased splenic CD4CD25FoxP3 cells, spleen siderosis, and increased marginal zones in the splenic white pulp were found. During AP, peripheral and splenic CD3(+) and CD3CD4 declined stronger in the p8(−/−) mice. The spleen of the p8(−/−) mice showed severe hypoplasia of the white pulp and mild hyperplasia of the red pulp. This was associated with a significantly increased rate of apoptosis. We conclude that p8 has no impact on the cellular composition of the adaptive and innate immune systems in noninflammatory conditions. However, it may limit apoptosis and maintain homeostasis of the immune reaction during AP.
Product Description
Supelco
Resorcinol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Resorcinol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Resorcinol, ≥98%, FG
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Sigma-Aldrich
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Sodium Thiosulfate Solution, 2 g/dL in deionized water
Sigma-Aldrich
Resorcinol, BioXtra, ≥99%
Hymecromone impurity A, European Pharmacopoeia (EP) Reference Standard
Resorcinol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Sodium thiosulfate, purum p.a., anhydrous, ≥98.0% (RT)
Sigma-Aldrich
Sodium thiosulfate, ReagentPlus®, 99%