Merck
  • Multifunctional disulfide-based cationic dextran conjugates for intravenous gene delivery targeting ovarian cancer cells.
Multifunctional disulfide-based cationic dextran conjugates for intravenous gene delivery targeting ovarian cancer cells.
Molecular pharmaceutics (2014-06-04)
Yanyan Song, Bo Lou, Peng Zhao, Chao Lin
ABSTRACT
A folate-decorated, disulfide-based cationic dextran conjugate having dextran as the main chain and disulfide-linked 1,4-bis(3-aminopropyl)piperazine (BAP) residues as the grafts was designed and successfully prepared as a multifunctional gene delivery vector for targeted gene delivery to ovarian cancer SKOV-3 cells in vitro and in vivo. Initially, a new bioreducible cationic polyamide (denoted as pSSBAP) was prepared by polycondensation reaction of bis(p-nitrophenyl)-3,3'-dithiodipropanoate, a disulfide-containing monomer, and BAP. It was found that the pSSBAP was highly efficient for in vitro gene delivery against MCF-7 and SKOV-3 cell lines. Subsequently, two cationic dextran conjugates with different amounts of BAP residues (denoted as Dex-SSBAP6 and Dex-SSBAP30, respectively) were synthesized by coupling BAP to disulfide-linked carboxylated dextran or coupling pSSBAP-oligomer to p-nitrophenyl carbonated dextran. Both two conjugates were able to bind DNA to form nanosized polyplexes with an improved colloidal stability in physiological conditions. The polyplexes, however, were rapidly dissociated to liberate DNA in a reducing environment. In vitro transfection experiments revealed that the polyplexes of Dex-SSBAP30 efficiently transfected SKOV-3 cells, yielding transfection efficiency that is comparable to that of linear polyethylenimine or lipofectamine 2000. AlamarBlue assay showed that the conjugates had low cytotoxicity in vitro at a high concentration of 100 mg/L. Further, Dex-SSBAP30 has primary amine side groups and thus allows for folate (FA) conjugation, yielding FA-coupled Dex-SSBAP30 (Dex-SSBAP30-FA). It was found that Dex-SSBAP30-FA was efficient for targeted gene delivery to SKOV-3 tumor xenografted in a nude mouse model by intravenous injection, inducing a higher level of gene expression in the tumor as compared to Dex-SSBAP30 lacking FA and comparable gene expression to linear polyethylenimine as one of the most efficient polymeric vectors for intravenous gene delivery in vivo. Disulfide-based cationic dextran system thus has a high potential for intravenous gene delivery toward cancer gene therapy.
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99.0%
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
1,4-Bis(3-aminopropyl)piperazine, ≥99%
Sigma-Aldrich
3,3′-Dithiodipropionic acid, 99%
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Dimethyl sulfoxide
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%