Merck
  • Specific ablation of Nampt in adult neural stem cells recapitulates their functional defects during aging.
Specific ablation of Nampt in adult neural stem cells recapitulates their functional defects during aging.
The EMBO journal (2014-05-09)
Liana R Stein, Shin-ichiro Imai
ABSTRACT
Neural stem/progenitor cell (NSPC) proliferation and self-renewal, as well as insult-induced differentiation, decrease markedly with age. The molecular mechanisms responsible for these declines remain unclear. Here, we show that levels of NAD(+) and nicotinamide phosphoribosyltransferase (Nampt), the rate-limiting enzyme in mammalian NAD(+) biosynthesis, decrease with age in the hippocampus. Ablation of Nampt in adult NSPCs reduced their pool and proliferation in vivo. The decrease in the NSPC pool during aging can be rescued by enhancing hippocampal NAD(+) levels. Nampt is the main source of NSPC NAD(+) levels and required for G1/S progression of the NSPC cell cycle. Nampt is also critical in oligodendrocytic lineage fate decisions through a mechanism mediated redundantly by Sirt1 and Sirt2. Ablation of Nampt in the adult NSPCs in vivo reduced NSPC-mediated oligodendrogenesis upon insult. These phenotypes recapitulate defects in NSPCs during aging, giving rise to the possibility that Nampt-mediated NAD(+) biosynthesis is a mediator of age-associated functional declines in NSPCs.
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, BioUltra, ≥99%
Sigma-Aldrich
Bis(cyclohexanone)oxaldihydrazone, ≥95% (TLC)
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
Anti-Nestin Antibody, clone rat-401, clone rat-401, Chemicon®, from mouse
Sigma-Aldrich
FK866 hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture
Sigma-Aldrich
AGK2, ≥97% (HPLC), powder
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse