Merck
Phosphopeptide analysis of rodent epididymal spermatozoa.
Journal of visualized experiments : JoVE (2015-01-16)
Mark A Baker, Louise Hetherington, Anita Weinberg, Tony Velkov
ABSTRACT
Spermatozoa are quite unique amongst cell types. Although produced in the testis, both nuclear gene transcription and translation are switched off once the pre-cursor round cell begins to elongate and differentiate into what is morphologically recognized as a spermatozoon. However, the spermatozoon is very immature, having no ability for motility or egg recognition. Both of these events occur once the spermatozoa transit a secondary organ known as the epididymis. During the ~12 day passage that it takes for a sperm cell to pass through the epididymis, post-translational modifications of existing proteins play a pivotal role in the maturation of the cell. One major facet of such is protein phosphorylation. In order to characterize phosphorylation events taking place during sperm maturation, both pure sperm cell populations and pre-fractionation of phosphopeptides must be established. Using back flushing techniques, a method for the isolation of pure spermatozoa of high quality and yield from the distal or caudal epididymides is outlined. The steps for solubilization, digestion, and pre-fractionation of sperm phosphopeptides through TiO2 affinity chromatography are explained. Once isolated, phosphopeptides can be injected into MS to identify both protein phosphorylation events on specific amino acid residues and quantify the levels of phosphorylation taking place during the sperm maturation processes.
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Titanium, wire reel, 200m, diameter 0.25mm, as drawn, 99.6+%
Titanium, wire reel, 20m, diameter 1.5mm, as drawn, 99.6+%
Titanium, wire reel, 5m, diameter 0.5mm, as drawn, 99.6+%
Titanium, wire reel, 5m, diameter 0.25mm, as drawn, 99.99+%
Titanium, tube, 500mm, outside diameter 2.03mm, inside diameter 1.55mm, wall thickness 0.24mm, annealed, 99.6+%
Titanium, wire reel, 1m, diameter 0.25mm, as drawn, 99.99+%
Titanium, wire reel, 20m, diameter 0.05mm, hard, 99.8%
Titanium, wire reel, 2m, diameter 1.0mm, as drawn, 99.99+%
Titanium, wire reel, 200m, diameter 0.125mm, annealed, 99.6+%
Titanium, wire reel, 5m, diameter 1.0mm, as drawn, 99.6+%
Titanium, wire reel, 25m, diameter 0.20mm, annealed, 99.6+%
Titanium, wire reel, 5m, diameter 0.75mm, as drawn, 99.6+%
Titanium, wire reel, 25m, diameter 1.0mm, as drawn, 99.6+%
Titanium, wire reel, 200m, diameter 0.20mm, annealed, 99.6+%