Merck
  • Adhesive strength of the luting technique for passively fitting screw-retained implant-supported prostheses: an in vitro evaluation.
Adhesive strength of the luting technique for passively fitting screw-retained implant-supported prostheses: an in vitro evaluation.
The International journal of prosthodontics (2015-01-15)
Maria Menini, Francesco Pera, Marco Migliorati, Paolo Pesce, Paolo Pera
ABSTRACT
This in vitro study evaluated the adhesive strength of a technique to lute implant cylinders to metal frameworks in implant-supported prostheses and ensure a good passive fit. Different height samples were tested: In group 1, implant cylinders were 5 mm long; in group 2, they were 10 mm long. A universal testing machine (Instron) was used to perform pullout tests. The luting technique provided enough adhesive strength for clinical use with greater adhesive strength in group 2 (mean pull-out strength: 2.85 kN in group 1 versus 3.79 kN in group 2). The luting technique provides enough adhesive strength for clinical use. Moreover, specimens with a larger surface for adhesion demonstrated higher adhesive strength compared with shorter specimens.
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Titanium, wire reel, 5m, diameter 0.75mm, as drawn, 99.6+%
Titanium, wire reel, 5m, diameter 1.0mm, as drawn, 99.6+%
Titanium, tube, 1000mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 25.4mm, inside diameter 23.62mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, tube, 1220mm, outside diameter 2.03mm, inside diameter 1.55mm, wall thickness 0.24mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 2.03mm, inside diameter 1.63mm, wall thickness 0.2mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 4.25mm, inside diameter 3.75mm, wall thickness 0.25mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 8.0mm, as drawn, 99.99+%
Titanium, rod, 200mm, diameter 9.5mm, annealed, 99.99+%
Titanium, rod, 200mm, diameter 16mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 25.4mm, inside diameter 23.62mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 2mm, as drawn, 99.99+%
Titanium, rod, 100mm, diameter 16mm, as drawn, 99.99+%
Titanium, rod, 500mm, diameter 4mm, annealed, 99.6+%
Titanium, rod, 500mm, diameter 6mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 9.5mm, inside diameter 8.2mm, wall thickness 0.65mm, annealed, 99.6+%
Titanium, rod, 25mm, diameter 50mm, 99.99+%
Titanium, tube, 1000mm, outside diameter 4.25mm, inside diameter 3.75mm, wall thickness 0.25mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 9.5mm, annealed, 99.99+%
Titanium, wire reel, 25m, diameter 0.20mm, annealed, 99.6+%