Merck
  • Diversification of 99Mo/99mTc separation: non–fission reactor production of 99Mo as a strategy for enhancing 99mTc availability.
Diversification of 99Mo/99mTc separation: non–fission reactor production of 99Mo as a strategy for enhancing 99mTc availability.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2014-12-30)
Maroor R A Pillai, Ashutosh Dash, Furn F Russ Knapp
ABSTRACT
This paper discusses the benefits of obtaining (99m)Tc from non-fission reactor-produced low-specific-activity (99)Mo. This scenario is based on establishing a diversified chain of facilities for the distribution of (99m)Tc separated from reactor-produced (99)Mo by (n,γ) activation of natural or enriched Mo. Such facilities have expected lower investments than required for the proposed chain of cyclotrons for the production of (99m)Tc. Facilities can receive and process reactor-irradiated Mo targets then used for extraction of (99m)Tc over a period of 2 wk, with 3 extractions on the same day. Estimates suggest that a center receiving 1.85 TBq (50 Ci) of (99)Mo once every 4 d can provide 1.48-3.33 TBq (40-90 Ci) of (99m)Tc daily. This model can use research reactors operating in the United States to supply current (99)Mo needs by applying natural (nat)Mo targets. (99)Mo production capacity can be enhanced by using (98)Mo-enriched targets. The proposed model reduces the loss of (99)Mo by decay and avoids proliferation as well as waste management issues associated with fission-produced (99)Mo.
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Molybdenum, foil, 150x150mm, thickness 0.125mm, annealed, 99.9%
Molybdenum, wire reel, 50m, diameter 0.5mm, annealed, 99.95%
Molybdenum, wire reel, 100m, diameter 0.03mm, hard, 99.95%
Molybdenum, wire reel, 2000m, diameter 0.035mm, as drawn, 99.95%
Molybdenum, foil, 100x100mm, thickness 0.15mm, annealed, 99.9%
Molybdenum, wire reel, 50m, diameter 0.125mm, annealed, 99.95%
Molybdenum, foil, 150x150mm, thickness 0.25mm, annealed, 99.9%
Molybdenum, wire reel, 2m, diameter 0.30mm, annealed, 99.95%
Molybdenum, wire reel, 10m, diameter 0.30mm, annealed, 99.95%
Molybdenum, wire reel, 2000m, diameter 0.03mm, hard, 99.95%
Molybdenum, wire reel, 100m, diameter 0.05mm, gold plated, 99.95%
Molybdenum, wire reel, 200m, diameter 0.2mm, as drawn, 99.95%
Molybdenum, wire reel, 5m, diameter 0.5mm, annealed, 99.95%
Molybdenum, wire reel, 100m, diameter 1.25mm, annealed, 99.95%
Molybdenum, wire reel, 50m, diameter 0.05mm, gold plated, 99.95%
Molybdenum, wire reel, 50m, diameter 0.25mm, annealed, 99.95%
Molybdenum, wire reel, 500m, diameter 0.025mm, hard, 99.95%
Molybdenum, wire reel, 5m, diameter 0.015mm, gold plated, 99.95%
Molybdenum, foil, 100x100mm, thickness 0.20mm, annealed, 99.9%
Molybdenum, foil, light tested, 25x25mm, thickness 0.05mm, annealed, 99.9%
Molybdenum, foil, not light tested, 25x25mm, thickness 0.005mm, 99.9%
Molybdenum, foil, light tested, 50x50mm, thickness 0.009mm, 99.9%
Molybdenum, foil, 6mm disks, thickness 0.007mm, 99.9%
Molybdenum, foil, 6mm disks, thickness 0.006mm, 99.9%