Merck
  • A novel point-of-use water treatment method by antimicrobial nanosilver textile material.
A novel point-of-use water treatment method by antimicrobial nanosilver textile material.
Journal of water and health (2014-12-05)
Hongjun Liu, Xiaosheng Tang, Qishan Liu
ABSTRACT
Pathogenic bacteria are one of the main reasons for worldwide water-borne disease causing a big threat to public health, hence there is an urgent need to develop cost-effective water treatment technologies. Nano-materials in point-of-use systems have recently attracted considerable research and commercial interests as they can overcome the drawbacks of traditional water treatment techniques. We have developed a new point-of-use water disinfection kit with nanosilver textile material. The silver nanoparticles were in-situ generated and immobilized onto cotton textile, followed by fixing to a plastic tube to make a water disinfection kit. By soaking and stirring the kit in water, pathogenic bacteria have been killed within minutes. The silver leaching from the kit was insignificant, with values <100 ppb - the current US EPA and WHO limit for silver level in drinking water. Herein, the nanosilver textile water disinfection kit could be a new, efficient and cost-effective point-of-use water treatment method for rural areas and emergency preparedness.
Sigma-Aldrich
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Silver, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Silver, tube, 500mm, outside diameter 1.1mm, inside diameter 0.75mm, wall thickness 0.175mm, as drawn, 99.95+%
Silver, tube, 500mm, outside diameter 7.0mm, inside diameter 6mm, wall thickness 0.5mm, as drawn, 99.95+%
Silver, wire reel, 5m, diameter 1.0mm, annealed, 99.99%
Silver, foil, not light tested, 150x150mm, thickness 0.008mm, 99.97%
Silver, insulated wire, 1m, conductor diameter 0.25mm, insulation thickness 0.005mm, polyurethane insulation, 99.99%
Silver, insulated wire, 10m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyester insulation, 99.99%
Silver, tube, 100mm, outside diameter 4.65mm, inside diameter 4.15mm, wall thickness 0.25mm, as drawn, 99.95+%
Silver, rod, 1000mm, diameter 3.5mm, as drawn, 99.95+%
Silver, insulated wire, 10m, conductor diameter 0.25mm, insulation thickness 0.023mm, polyester insulation, 99.99%
Silver, insulated wire, 20m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Silver, rod, 100mm, diameter 5.0mm, as drawn, 99.95+%
Silver, insulated wire, 20m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyester insulation, 99.99%
Silver, insulated wire, 10m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyimide insulation, 99.9%
Silver, tube, 100mm, outside diameter 3.5mm, inside diameter 3.04mm, wall thickness 0.23mm, as drawn, 99.95+%
Silver, insulated wire, 10m, conductor diameter 0.25mm, insulation thickness 0.015mm, epoxy insulation, 99.99%
Silver, sphere, 50pcs, diameter 0.5mm, precision sphere grade
Silver, tube, 100mm, outside diameter 4.65mm, inside diameter 4.23mm, wall thickness 0.21mm, as drawn, 99.95+%
Silver, tube, 1000mm, outside diameter 6.0mm, inside diameter 4mm, wall thickness 1.0mm, as drawn, 99.95+%
Silver, tube, 200mm, outside diameter 2.8mm, inside diameter 2.5mm, wall thickness 0.15mm, as drawn, 99.95+%
Silver, insulated wire, 50m, conductor diameter 0.25mm, insulation thickness 0.023mm, polyester insulation, 99.99%
Silver, rod, 50mm, diameter 8.0mm, as drawn, 99.95+%
Silver, insulated wire, 5m, conductor diameter 0.075mm, insulation thickness 0.010mm, nylon insulation, 99.99%
Silver, rod, 500mm, diameter 2.0mm, as drawn, 99.95+%