Merck
  • Primary squamous cell carcinoma of the duodenum effectively treated with TS-1: a case report.
Primary squamous cell carcinoma of the duodenum effectively treated with TS-1: a case report.
Nihon Shokakibyo Gakkai zasshi = The Japanese journal of gastro-enterology (2014-12-09)
Toshihiro Fujita, Yuichiro Nasu, Hitomi Hamamoto, Fumiko Miyata, Akihiko Oshige, Shuho Shigenobu, Shuji Kanmura, Masatsugu Numata, Shinichiro Aozaki, Akio Ido
ABSTRACT
An 89-year-old man was admitted to our hospital for thorough investigation of refractory diabetes mellitus, which revealed primary squamous cell carcinoma of the duodenum. After two courses of chemotherapy, follow-up esophagoduodenogastroscopy and duodenal biopsy showed no evidence of tumor. No findings were suggestive of recurrence of the primary lesion 19 months after starting chemotherapy. This case suggests that chemotherapy including TS-1 may be effective for treating unresectable primary squamous cell carcinoma of the duodenum.
Product Description
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Titanium, rod, 200mm, diameter 2mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 16mm, as drawn, 99.99+%
Titanium, microfoil, disks, 10mm, thinness 1.0μm, specific density 429μg/cm2, permanent mylar 3.5μm support, 99.6+%
Titanium, rod, 490mm, diameter 2mm, annealed, 99.6+%
Titanium, tube, 1220mm, outside diameter 2.03mm, inside diameter 1.55mm, wall thickness 0.24mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 25mm, diameter 16mm, as drawn, 99.99+%
Titanium, rod, 500mm, diameter 1.5mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 6mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 25.4mm, inside diameter 23.62mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 5mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 1.6mm, inside diameter 1.2mm, wall thickness 0.2mm, hard, 99.6+%
Titanium, mesh, 100x100mm, nominal aperture 0.19mm, wire diameter 0.23mm, 60x60 wires/inch, open area 20%, twill weave
Titanium, rod, 100mm, diameter 2mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 9.5mm, annealed, 99.99+%
Titanium, rod, 100mm, diameter 2mm, as drawn, 99.99+%
Titanium, rod, 10mm, diameter 50mm, 99.99+%
Titanium, rod, 1000mm, diameter 3.0mm, annealed, 99.6+%
Titanium, microfoil, disks, 25mm, thinness 0.5μm, specific density 225.4μg/cm2, permanent mylar 3.5μm support, 99.6+%
Titanium, rod, 100mm, diameter 5mm, annealed, 99.6+%